missing translation for 'onlineSavingsMsg'
Learn More

Bromaminic acid, 90+%

Product Code. 15474787
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474787 100 g 100g
15464787 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474787 Supplier Thermo Scientific Alfa Aesar Supplier No. H32110.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 116-81-4 | C14H7BrNNaO5S | 404.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-81-4
Molecular Formula C14H7BrNNaO5S
Molecular Weight (g/mol) 404.17
MDL Number MFCD00035694
InChI Key TXMRAEGWZZVGIH-UHFFFAOYSA-M
Synonym bromaminic acid, bromamine acid, 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid, alizarine cyanol grey g, unii-sbz7fun4bk, 1-amino-4-bromoanthraquinone-2-sulphonic acid, 1-amino-4-bromoanthraquinone-2-sulfonic acid, sbz7fun4bk, 1-amino-4-bromo-9,10-dioxo-9,10-dihydro-anthracene-2-sulfonic acid, 2-anthracenesulfonic acid, 1-amino-4-bromo-9,10-dihydro-9,10-dioxo
PubChem CID 22628
IUPAC Name 1-amino-4-bromo-9,10-dioxoanthracene-2-sulfonic acid
SMILES [Na+].NC1=C(C=C(Br)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O

Specifications

Melting Point ∽280°C (decomposition)
Odor Odorless
Quantity 100 g
Formula Weight 382.19
Percent Purity ≥90%
Chemical Name or Material Bromaminic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.