missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine hydrochloride, 98+%, Thermo Scientific™

A mucolytic agent

140.00 € - 419.00 €

Chemical Identifiers

CAS 611-75-6
Molecular Formula C14H21Br2ClN2
Molecular Weight (g/mol) 412.59
MDL Number MFCD00056626
InChI Key SXIVGYTWSVJOLM-UHFFFAOYNA-N
Synonym bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
PubChem CID 5702220
ChEBI CHEBI:31303
IUPAC Name hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
SMILES [H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15495689
SDS View Documents
Thermo Scientific Alfa Aesar
J63451.22
100g
140.00 €
100g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15405699
SDS View Documents
Thermo Scientific Alfa Aesar
J63451.36
500g
419.00 €
500g
View alternative products
Save Up to 
»
Estimated Shipment: 23-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

611-75-6
412.59
SXIVGYTWSVJOLM-UHFFFAOYNA-N
5702220
hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
C14H21Br2ClN2
MFCD00056626
bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
CHEBI:31303
[H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N

Specifications

611-75-6
MFCD00056626
14,1391
SXIVGYTWSVJOLM-UHFFFAOYNA-N
hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
5702220
412.6
Bromhexine hydrochloride
C14H21Br2ClN2
4848376
bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
[H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N
412.59
CHEBI:31303
≥98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 210-280-8

missing translation for 'rtecsNumber' : XS9950000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.