missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine hydrochloride, 98+%, Thermo Scientific™

A mucolytic agent

Brand:  Thermo Scientific Alfa Aesar J63451.22

Quantity 100g

 View more versions of this product

Product Code. 15495689

  • 140.00 € / 100g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

611-75-6
412.59
SXIVGYTWSVJOLM-UHFFFAOYNA-N
5702220
hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
C14H21Br2ClN2
MFCD00056626
bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
CHEBI:31303
[H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N

Specifications

611-75-6
MFCD00056626
4848376
bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
[H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N
412.59
CHEBI:31303
≥98%
C14H21Br2ClN2
100g
14,1391
SXIVGYTWSVJOLM-UHFFFAOYNA-N
hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
5702220
412.6
Bromhexine hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.