missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| CAS | 7732-18-5, 7758-02-3, 7758-01-2 |
| Quantity | 1 L |
| Molecular Formula | H2O |
| UN Number | NONH for all modes of transport |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| SMILES | O |
| IUPAC Name | water |
| Molecular Weight (g/mol) | 18.02 |
| Concentration or Composition (by Analyte or Components) | 0.05M |
| Chemical Name or Material | Bromide bromate Standard Solution |
Safety and Handling
Product Identifier
- bromide bromate standard solution
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: potassium bromate, potassium bromide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?