missing translation for 'onlineSavingsMsg'
Learn More

Bromine (Bromide-Bromate) Solution, Honeywell Fluka™

Product Code. 15663520 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
2 L
Unit Size:
2 L
500mL
This item is not returnable. View return policy

Product Code. 15663520

Brand: Honeywell-Fluka 3186982L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, 0.05 M Br2 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00143870

Specifications

Quantity 2 L
UN Number NONH for all modes of transport
Density 1.005g/mL at 25°C
Chemical Name or Material Bromine (Bromide-Bromate) Solution
compliance-icons
Product Identifier
  • Bromine (bromide-bromate) solution
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium bromate, potassium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.