missing translation for 'onlineSavingsMsg'
Learn More

Bromine concentrate, For 1L standard solution, 0.05 M Br2, Solstice

Product Code. 15655190
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15655190 1 Pc. Each
15665190 6 x 1 Ea. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15655190 Supplier Honeywell-Fluka Supplier No. 380401EA
  • ONLINE EXCLUSIVE PRICE
    100.00 €
    93.93 € / Pack of 1
    Save 6.07 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 1L standard solution, 0.05 M Br2

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
IUPAC Name molecular bromine
SMILES [Br-]

Specifications

Quantity 1 Pc.
MDL Number MFCD00010896
UN Number NONH for all modes of transport
Formula Weight 159.81g/mol
Density 1.307g/mL at 25°C
Chemical Name or Material Bromine concentrate
compliance-icons
Product Identifier
  • Bromine concentrate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: potassium bromate, Potassium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.