missing translation for 'onlineSavingsMsg'
Learn More

Bromine, Extra Pure, SLR, Fisher Chemical™

Product Code. 10510721 Shop All Fisher Chemical Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
missing translation for 'unitSize':
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
10510721 100 mL 100mL
10182180 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10510721 missing translation for 'mfr' Fisher Chemical missing translation for 'supplierNo' B/4000/PB05
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number 10896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
IUPAC Name bromide
SMILES [Br-]

Specifications

Boiling Point 58.7°C
Melting Point -7.2°C
Color Brown-Red
Physical Form Liquid
Quantity 100 mL
Formula Weight 159.82g/mol
Packaging Plastic Coated Glass bottle
Vapor Pressure 230mbar at 20°C
Density Vapor Density: 5.51
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.