missing translation for 'onlineSavingsMsg'
Learn More

Bromine Water, Saturated, Chem-Lab

Product Code. 13278293
Change view
Click to view available options
Quantity:
1 L
Packaging:
GVB
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13278293 1 L GVB 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13278293 Supplier Chem-Lab Supplier No. CL02.0211.1000
  • ONLINE EXCLUSIVE PRICE
    99.70 €
    57.50 € / 1 L
    Save 42.20 €
    42% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Saturated bromine solution for laboratory use, ASTM-D-129

Specifications

CAS 7726-95-6
Molecular Formula Br
UN Number UN1744
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES [Br-]
IUPAC Name molecular bromine
Molecular Weight (g/mol) 79.91
PubChem CID 24408
ChEBI CHEBI:29224
Formula Weight 159.81g/mol
Packaging GVB
Density 1.02g/mL
Quantity 1 L
Chemical Name or Material Bromine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromine water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Bromine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.