missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid, 98+%

Product Code. 11487017
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487017 5000 g 5000g
11477017 250 g 250g
11467017 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487017 Supplier Thermo Scientific Alfa Aesar Supplier No. A14403.0I
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 79-08-3 | C2H3BrO2 | 138.948 g/mol

Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water, alcohol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to light and air. Handle and store under inert gas. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-08-3
Molecular Formula C2H3BrO2
Molecular Weight (g/mol) 138.948
MDL Number MFCD00002678
InChI Key KDPAWGWELVVRCH-UHFFFAOYSA-N
Synonym bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo
PubChem CID 6227
IUPAC Name 2-bromoacetic acid
SMILES C(C(=O)O)Br

Specifications

Melting Point 44°C to 49°C
Density 1.93
Boiling Point 207°C to 209°C
Flash Point >110°C (230°F)
Odor Pungent
Linear Formula BrCH2CO2H
Quantity 5000 g
UN Number UN3425
Beilstein 506167
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,1400
Solubility Information Soluble in water, alcohol
Formula Weight 138.95
Percent Purity ≥98%
Chemical Name or Material Bromoacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.