missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid, 98+%

Product Code. 10216883 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10216883 2.5 kg Glass bottle 2.5kg
10316360 250 g Glass bottle 250g
10112420 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10216883 Supplier Thermo Scientific Acros Supplier No. 106570025
  • ONLINE EXCLUSIVE PRICE
    181.00 €
    167.00 € / 2.5kg
    Save 14.00 €
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-08-3 | C2H3BrO2 | 138.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-08-3
Molecular Formula C2H3BrO2
Molecular Weight (g/mol) 138.95
MDL Number MFCD00002678
InChI Key KDPAWGWELVVRCH-UHFFFAOYSA-N
Synonym bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo
PubChem CID 6227
IUPAC Name 2-bromoacetic acid
SMILES C(C(=O)O)Br

Specifications

Melting Point 49.0°C to 51.0°C
Color White to Yellow
Density 1.9300g/mL
Boiling Point 208.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrCH2CO2H
Quantity 2.5 kg
Beilstein 02,213
Merck Index 15,1407
Specific Gravity 1.93
Solubility Information Solubility in water: 900g/100mL (25°C). Other solubilities:g/100g(25°C): methanol-900,ether-735,benzene-308,carbon tetrachloride-168,soluble in acetone and alcohol,practically insoluble in n-heptane
Formula Weight 138.95
Percent Purity 99%
Physical Form Crystalline Mass or Crystalline Powder, Flakes, Crystals and/or Chunks
Chemical Name or Material Bromoacetic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromoacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Bromoacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.