missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic anhydride, 95%

Product Code. 10244884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10244884 5 g Glass bottle 5g
10788425 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10244884 Supplier Thermo Scientific Acros Supplier No. 426700050
  • ONLINE EXCLUSIVE PRICE
    152.00 €
    147.00 € / 5g
    Save 5.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13094-51-4 | C4H4Br2O3 | 259.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13094-51-4
Molecular Formula C4H4Br2O3
Molecular Weight (g/mol) 259.87
MDL Number MFCD00045041
InChI Key FUKOTTQGWQVMQB-UHFFFAOYSA-N
Synonym bromoacetic anhydride, 2-bromoacetic anhydride, acetic acid, 2-bromo-,1,1'-anhydride, 2-bromoacetyl 2-bromoacetate, bromoaceticanhydride, acetic acid, bromo-, anhydride
PubChem CID 4259817
IUPAC Name (2-bromoacetyl) 2-bromoacetate
SMILES C(C(=O)OC(=O)CBr)Br

Specifications

Melting Point 40°C to 44°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ex Br) (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2C(=O)OC(=O)CH2Br
Quantity 5 g
Formula Weight 259.87
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Bromoacetic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.