missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetonitrile, 96%

Product Code. 11461658
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461658 10 g 10g
11471658 50 g 50g
11481658 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461658 Supplier Thermo Scientific Alfa Aesar Supplier No. A13933.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-17-0 | C2H2BrN | 119.95 g/mol

Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 590-17-0
Molecular Formula C2H2BrN
Molecular Weight (g/mol) 119.95
MDL Number MFCD00001884
InChI Key REXUYBKPWIPONM-UHFFFAOYSA-N
Synonym bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile
PubChem CID 11534
IUPAC Name 2-bromoacetonitrile
SMILES BrCC#N

Tekniske data

Density 1.722
Boiling Point 150°C
Flash Point 81°C (178°F)
Refractive Index 1.48
Quantity 10 g
UN Number UN3276
Beilstein 956569
Solubility Information Insoluble in water.
Formula Weight 119.95
Percent Purity 96%
Chemical Name or Material Bromoacetonitrile
Vis mere Vis mindre
compliance-icons compliance-icons
Produktidentifikator
  • Bromoacetonitrile
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faresætning
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Bromoacetonitrile

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.