missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl bromide, 98%

Product Code. 10336360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10336360 500 mL Glass bottle 500mL
10448820 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336360 Supplier Thermo Scientific Acros Supplier No. 106605000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 598-21-0 | C2H2Br2O | 201.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-21-0
Molecular Formula C2H2Br2O
Molecular Weight (g/mol) 201.85
MDL Number MFCD00000115
InChI Key LSTRKXWIZZZYAS-UHFFFAOYSA-N
Synonym bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
PubChem CID 61147
IUPAC Name 2-bromoacetyl bromide
SMILES C(C(=O)Br)Br

Specifications

Color White
Density 2.3100g/mL
Boiling Point 147°C to 150°C
Flash Point >105°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2COBr
Refractive Index 1.5450 to 1.547
Quantity 500 mL
Beilstein 02, 215
Specific Gravity 2.31
Solubility Information Solubility in water: reacts. Other solubilities: solulbe in acetone,carbontetrachloride
Formula Weight 201.85
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bromoacetyl bromide, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Bromoacetyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.