missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl bromide, 98%

Product Code. 11305279
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11305279 500 g 500g
11385269 25 g 25g
11395269 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11305279 Supplier Thermo Scientific Alfa Aesar Supplier No. A19580.36
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 598-21-0 | C2H2Br2O | 201.845 g/mol

Bromoacetyl bromide is widely used in the synthesis of fine chemicals, agrochemicals, dyes and pharmaceuticals. As an acylating agent, it reacts with 10H-phenothiazine to give 10-bromoacetyl-10H-phenothiazine. Similarly, it is also involved in the preparation of bromoactylated starch which is a biodegradable thermoplastic copolymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetyl bromide is widely used in the synthesis of fine chemicals, agrochemicals, dyes and pharmaceuticals. As an acylating agent, it reacts with 10H-phenothiazine to give 10-bromoacetyl-10H-phenothiazine. Similarly, it is also involved in the preparation of bromoactylated starch which is a biodegradable thermoplastic copolymer.

Solubility
Immiscible with water.

Notes
Moisture Sensitive. Store in cool place. Hydrolyses readily. Incompatible with strong oxidizing agents, alcohols and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-21-0
Molecular Formula C2H2Br2O
Molecular Weight (g/mol) 201.845
MDL Number MFCD00000115
InChI Key LSTRKXWIZZZYAS-UHFFFAOYSA-N
Synonym bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
PubChem CID 61147
IUPAC Name 2-bromoacetyl bromide
SMILES C(C(=O)Br)Br

Specifications

Density 2.324
Boiling Point 147°C to 150°C
Flash Point >105°C (221°F)
Odor Acrid
Refractive Index 1.546
Quantity 500 g
UN Number UN2513
Beilstein 605440
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 201.86
Percent Purity 98%
Chemical Name or Material Bromoacetyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.