missing translation for 'onlineSavingsMsg'
Learn More

Bromobenzene-d5, for NMR, 99+% atom D, ACROS Organics™

Product Code. 10133810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass Bottle
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133810 5 mL Glass Bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10133810 Supplier Thermo Scientific Acros Supplier No. 174730050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Physical Form Liquid
Color Colorless
Deuterium (D) 99.0% min.
Packaging Glass Bottle
Water 0.1% max. (Coulometric)
Quantity 5 mL
CAS 4165-57-5
Molecular Formula C6H5Br
Linear Formula C6D5Br
MDL Number MFCD00000056
Beilstein 05, III, 561
Synonym bromobenzene-d5, bromo 2h5 benzene, 1-bromo 2 h? benzene, bromobenzene d5, benzene-d5, bromo, 1-bromo 2h5 benzene, c6d5br, bromobenzene-d5, 99.5 atom % d, bromobenzene-d5, reagent grade, >=99 atom % d cp
InChI Key QARVLSVVCXYDNA-RALIUCGRSA-N
SMILES [2H]C1=C([2H])C([2H])=C(Br)C([2H])=C1[2H]
IUPAC Name 1-bromo(²H₅)benzene
Molecular Weight (g/mol) 162.04
PubChem CID 2723968
Formula Weight 162.06
Grade NMR
Chemical Name or Material Bromobenzene-d5, 99+ atom % D
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.