Learn More
R&D Systems™ BromoCatch™ GFP Expression Plasmid (N-term, pCMV, Puro)
Product Code. 31900333
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity unitSize
31900333 50 μg 50µg
1 options
Product Code. 31900333 Supplier R&D Systems™ Supplier No. RDEHBC01GFP050
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This plasmid enables expression of proteins fused to the BromoCatch™ tag, a covalent self-labeling tag derived from a BET bromodomain.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 μg