Learn More
Thermo Scientific Chemicals Bromocresol Green
Description
Bromocresol Green is a pH sensitive indicator used in the growth medium for microorganisms and titrations. It is useful in various colorimetric detection studies and as an actively tracking dye for DNA agarose gel electrophoresis. It acts as an inhibitor of prostaglandin E2 transport protein. It is also used in TLC for visualizing compounds containing functional groups having pKa less than 5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromocresol Green is a pH sensitive indicator used in the growth medium for microorganisms and titrations. It is useful in various colorimetric detection studies and as an actively tracking dye for DNA agarose gel electrophoresis. It acts as an inhibitor of prostaglandin E2 transport protein. It is also used in TLC for visualizing compounds containing functional groups having pKa less than 5.
Solubility
Soluble in benzene, ethanol and diethyl ether. Sparingly soluble in water.
Notes
Transition interval: pH 3.8 (yellow) to pH 5.4 (blue).Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼225°C (decomposition) |
| Beilstein | 372527 |
| Merck Index | 14,1386 |
| Solubility Information | Soluble in benzene,ethanol and diethyl ether. Sparingly soluble in water. |
| Quantity | 25 g |
| Formula Weight | 698.02 |
| Chemical Name or Material | Bromocresol Green |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.