missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, ACS

Product Code. 15416799
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416799 25 g 25g
15406799 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416799 Supplier Thermo Scientific Chemicals Supplier No. J65225.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol

Bromocresol Green is used in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualization of the compounds with functional groups whose pKa is below 5.0. And it is ready-to-use spray and dip reagent for chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green is used in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualization of the compounds with functional groups whose pKa is below 5.0. And it is ready-to-use spray and dip reagent for chromatography.

Solubility
Soluble in water, ethanol, methyl cellosolve.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005874
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

Melting Point 225°C (decomposition)
Beilstein 372527
Merck Index 14,1386
Color Brown to Yellow
Solubility Information Soluble in water,ethanol,methyl cellosolve.
Quantity 25 g
Physical Form Powder
Formula Weight 698.04
Grade ACS
Chemical Name or Material Bromocresol Green

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.