missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol green indicator, Reag. Ph. Eur., Solstice

Product Code. 15693840
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15693840 25 g Glass Bottle 25g
15603850 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15693840 Supplier Honeywell-Fluka Supplier No. 3274225G
  • ONLINE EXCLUSIVE PRICE
    312.00 €
    262.57 € / 25g
    Save 49.43 €
    15% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005875
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym 3′,3′′,5′,5′′-Tetrabromo-m-cresolsulfonephthalein, Bromocresol Blue
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

CAS Max % 100.0000%
Packaging Glass Bottle
Linear Formula C19H10Br4O5S
Beilstein 372527
Color Brown
Quantity 25 g
Formula Weight 698.01
Percent Purity 100%
Grade Ph. Eur.
Chemical Name or Material Bromocresol green indicator, Reag. Ph. Eur.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.