missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, pure, indicator grade

Product Code. 10498870 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10498870

Brand: Thermo Scientific Chemicals 151310250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bromocresol Green, C21H14Br4O5S, CAS Number-76-60-8, 3', 3″, 5', 3',3″,5',5″-Tetrabromo-m-cresolsulfonephthalein, BCG, 5g, Brown, 200-972-8, 698.01, Pure, Warning, GHS H Statement: May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. | CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005874
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5'
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

Merck Index 15, 1393
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in benzene, ethanol (30g/L), very soluble in diethyl ether and ethyl acetate
Infrared Spectrum Authentic
Formula Weight 698.01
Grade Pure
Packaging Glass bottle
Specific Extinction 600 min. (617nm, in buffer)
Visual Transition Interval from pH 3.8 (yellow) to pH 5.4 (blue)
Color Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bromocresol Green
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.