missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, sodium salt, ACS reagent

Product Code. 10005760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10005760 5 g Glass bottle 5g
10566592 10 g Glass bottle 10g
10075013 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10005760 Supplier Thermo Scientific Chemicals Supplier No. 402940050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 67763-24-0 | C21H13Br4NaO5S | 720.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67763-24-0
Molecular Formula C21H13Br4NaO5S
Molecular Weight (g/mol) 720.00
MDL Number MFCD00148898
InChI Key HEFSGAHJDGZCHA-UHFFFAOYNA-M
Synonym 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein sodium salt
PubChem CID 23675901
IUPAC Name sodium;2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenolate
SMILES [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C

Specifications

Clarity Passes Test
Infrared Spectrum Authentic
Formula Weight 720
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 3.8 (yellow) to pH 5.4 (blue)
Color Black to Brown/Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Bromocresol Green, sodium salt, Pure

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.