missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green sodium salt, ACS

Product Code. 15486299
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486299 25 g 25g
15476299 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486299 Supplier Thermo Scientific Chemicals Supplier No. J64284.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62625-32-5 | C21H13Br4NaO5S

Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

Solubility
40mg/ml in water; 60mg/ml in ethanolSoluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-32-5
Molecular Formula C21H13Br4NaO5S
MDL Number MFCD00148898
Synonym Bromocresol Green, water soluble

Specifications

Melting Point 230°C (decomposition)
Merck Index 14,1386
Color Brown to Red
Solubility Information 40mg/ml in water; 60mg/ml in ethanolSoluble in water and ethanol.
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 720.02
Grade ACS
Chemical Name or Material Bromocresol Green sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.