missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Sodium Salt, ACS, Thermo Scientific™

Product Code. 15835098 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15835098 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15835098 Supplier Thermo Scientific Supplier No. J12355.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as tracking dye in alkaline agarose gel electrophoresis

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point ∼230°C (decomposition)
CAS 62625-32-5
Molecular Formula C21H13Br4O5SNa
MDL Number MFCD00148898
Formula Weight 720
Grade ACS
Packaging Plastic bottle
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bromocresol Green sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.