missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Solution, Honeywell™ Fluka™

Product Code. 15620580 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15620580 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15620580 Supplier Honeywell-Fluka Supplier No. 02559500ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indicator, ready-to-use

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 89.5deg.C
CAS 76-60-8
Molecular Formula C21H14Br4O5S
MDL Number MFCD00005874
UN Number UN1219
Beilstein 372527
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
Molecular Weight (g/mol) 698.014
PubChem CID 6451
Formula Weight 698.01g/mol
Packaging Glass bottle
Density 0.79g/mL at 20°C
Quantity 500 mL
Flash Point 12°C (53.6°F)
Chemical Name or Material Bromocresol Green Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.