missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, sodium salt, pure, water soluble, indicator

Product Code. 10337660 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10337660

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67763-24-0 | C21H13Br4NaO5S | 720.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67763-24-0
Molecular Formula C21H13Br4NaO5S
Molecular Weight (g/mol) 720.00
MDL Number MFCD00148898
InChI Key HEFSGAHJDGZCHA-UHFFFAOYNA-M
Synonym 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein sodium salt
PubChem CID 23675901
IUPAC Name sodium;2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenolate
SMILES [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C

Specifications

Clarity (0.1% in water) clear
Infrared Spectrum Authentic
Formula Weight 720
Grade Extra Pure
Packaging Glass bottle
UV Spectrometry Complies
Visual Transition Interval from pH 3.8 (yellow) to pH 5.4 (blue)
Color Black/Green to Brown
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Bromocresol Green, sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.