missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple Indicator, Reag. Ph. Eur., Solstice

Product Code. 15673820
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
Unit Size:
1kg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15673820 1 kg Glass Bottle 1kg
15693820 5 g Glass Bottle 5g
15683820 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15673820 Supplier Honeywell-Fluka Supplier No. 326421KG
  • ONLINE EXCLUSIVE PRICE
    1565.00 €
    1314.20 € / 1kg
    Save 250.80 €
    16% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
MDL Number MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
PubChem CID 8273
ChEBI CHEBI:86154
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Melting Point 240°C
Packaging Glass Bottle
Suitability Complies for change range
UN Number NONH for all modes of transport
Beilstein 359618
Color Dark Purple
pH 5.2 to 6.8 (yellow to purple)
Quantity 1 kg
Formula Weight 540.22g/mol
Grade Indicator
Chemical Name or Material Bromocresol Purple, Reag. Ph. Eur.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.