missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Purple Indicator, Reag. Ph. Eur., Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
Unit Size:
1kg
25g
5g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 240°C |
| UN Number | NONH for all modes of transport |
| Beilstein | 359618 |
| Formula Weight | 540.22g/mol |
| Packaging | Glass Bottle |
| Suitability | Complies for change range |
| pH | 5.2 to 6.8 (yellow to purple) |
| Quantity | 5 g |
| Chemical Name or Material | Bromocresol Purple, Reag. Ph. Eur. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction