missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Purple, pure, indicator grade

Product Code. 10295570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
140 g
1 kg
Packaging:
Glass bottle
Unit Size:
140g
1kg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10295570 5 g Glass bottle 5g
10347660 25 g Glass bottle 25g
15760809 140 g Glass bottle 140g
11926801 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10295570 Supplier Thermo Scientific Chemicals Supplier No. 151330050
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 115-40-2 | C21H16Br2O5S | 540.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
MDL Number MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
Synonym Bromcresol Purple, BCP, 5', 5''-Dibromo-o-cresolsulfonephthalein
PubChem CID 8273
ChEBI CHEBI:86154
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Absorbance (20 mg/L in methanol) Complies, (20 mg/L in methanol) Complies at 0nm
Melting Point 245.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Visual Transition Interval from pH 5.2 (yellow) to pH 6.8 (purple)
Color Pink or Salmon
Solubility Information Solubility in water: practically insoluble. Other solubilities: 75g/L in ethanol (20°C)
Quantity 5 g
Physical Form Powder
IUPAC Name 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Formula Weight 540.21
Grade Pure
Chemical Name or Material Bromocresol Purple
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.