missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple Sodium Salt, MP Biomedicals™

Product Code. 11487781 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487781 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11487781 Supplier MP Biomedicals Supplier No. 0215051805
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Stains and dyes

Specifications

CAS 62625-30-3
Molecular Formula C21H15Br2NaO5S
MDL Number MFCD00148896
Synonym 5, 5-Dibromo-o-cresolsulfonphthalein sodium salt
InChI Key KVSJYIXUWNOWBD-UGAWPWHASA-M
SMILES CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]
IUPAC Name sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol) 562.204
PubChem CID 23665568
Formula Weight 562.22
Color Orange
Quantity 5 g
Physical Form Powder
Chemical Name or Material Bromocresol Purple Sodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.