missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Solution, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11484669 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484669 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484669 Supplier Reagecon Supplier No. 1012601
  • ONLINE EXCLUSIVE PRICE
    73.50 €
    50.24 € / 100mL
    Save 23.26 €
    31% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a pH indicator and tracking dye. Bromocresol Green Solution, EP Grade, Reagecon™ is a ready-to-use analytical volumetric solution used in manual and automatic titrations. It is manufactured according to European Pharmacopoeia Chapter 4 (4.1.1) and prepared in an ISO 9001 facility.

Bromocresol green (BCG) is a dye of the triphenylmethane family. It is used as a pH indicator in applications such as growth mediums for microorganisms and titrations. It is also used as a tracking dye for DNA agarose gel electrophoresis. It functions as an inhibitor of the prostaglandin E2 transport protein.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use reagent eliminates the time and labor of preparing precise volumetric solutions

Specifications

Melting Point 89°C
CAS 76-60-8
Molecular Formula C21H14Br4O5S
Synonym BCG
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
Molecular Weight (g/mol) 698.014
PubChem CID 6451
Formula Weight 698.01g/mol
Density 0.839g/cm3
Color Green
pH 3.8 to 5.4
Quantity 100 mL
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • BROMOCRESOL GREEN SOLUTION
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.