missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclohexane, 98%

Product Code. 11414673
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414673 2500 g 2500g
11494663 100 g 100g
11404673 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414673 Supplier Thermo Scientific Alfa Aesar Supplier No. A11593.0E
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 108-85-0 | C6H11Br | 163.06 g/mol

Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air & Light Sensitive. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-85-0
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.06
MDL Number MFCD00003819
InChI Key AQNQQHJNRPDOQV-UHFFFAOYSA-N
Synonym cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n
PubChem CID 7960
IUPAC Name bromocyclohexane
SMILES BrC1CCCCC1

Specifications

Melting Point -57°C
Density 1.335
Boiling Point 166°C to 167°C
Flash Point 62°C (143°F)
Refractive Index 1.495
Quantity 2500 g
Beilstein 1098373
Sensitivity Light sensitive
Merck Index 14,2730
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 163.06
Percent Purity 98%
Chemical Name or Material Bromocyclohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.