missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, 96%, stabilized

Product Code. 13468378 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
2.5 L
250mL
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13468378 5 L Plastic drum 5 L
10256110 250 mL Glass bottle 250mL
10377570 1 L Glass bottle 1 L
10573244 2.5 L Plastic drum 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13468378 Supplier Thermo Scientific Acros Supplier No. 158200050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-25-2 | CHBr3 | 252.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-25-2 , 64-17-5
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.73
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br

Specifications

Melting Point 8.0°C
Color Colorless to Orange-Yellow
Density 2.6300g/mL
Boiling Point 150.0°C to 151.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic drum
Linear Formula CHBr3
Quantity 5 L
Beilstein 01,68
Fieser 12,74
Merck Index 15,143
Specific Gravity 2.63
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,benzene,chloroform,ether,,petroleum ether,acetone and oils
Stabilizer 3 to 4 wt% ethanol
Formula Weight 252.73
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Bromoform
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.