missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, 99+%, stabilized

Product Code. 10331274 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10331274 100 mL Glass bottle 100mL
10743221 25 mL Glass bottle 25mL
10687094 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10331274 Supplier Thermo Scientific Acros Supplier No. 220691000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-25-2 | CHBr3 | 252.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-25-2 , 64-17-5
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.73
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br

Specifications

Melting Point 8°C
Color White
Density 2.8900g/mL
Boiling Point 150°C to 151°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (excluding stabilizer) (GC)
Packaging Glass bottle
Linear Formula CHBr3
Quantity 100 mL
Beilstein 01, 68
Fieser 12,74
Merck Index 15, 1430
Specific Gravity 2.89
Solubility Information Solubility in water: slightly soluble
Stabilizer 1.5 wt% max. ethanol
Formula Weight 252.73
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Bromoform
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.