missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoform-d, CDN
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Bromoform-d |
| CAS | 2909-52-6 |
| Molecular Formula | C D Br3 |
| Quantity | 10 g |
| Synonym | Methane-d, tribromo- (7 CI, 8 CI, 9 CI), Bromoform-d (6 CI), Bromoform-d1, Deuteriobromoform, Deuteriotribromomethane, Deuterobromoform, Deuterotribromomethane, Tribromodeuteriomethane, Tribromodeuteromethane, Tribromomethane-d, Bromoform-d1, Bromoform-D |
| InChI Formula | InChI=1 S/CHBr3/c2-1(3)4/h1H/i1D |
| SMILES | [2 H]C(Br)(Br)Br |
| IUPAC Name | tribromo(deuterio)methane |
| Molecular Weight (g/mol) | 253.7368 |
| Formula Weight | 250.7691 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction