missing translation for 'onlineSavingsMsg'
Learn More

Bromopentacarbonylmanganese(I), 98%

Product Code. 11333728
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11333728

Brand: Thermo Scientific Alfa Aesar 039310.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14516-54-2 | C5BrMnO5- | 274.892 g/mol

Bromopentacarbonylmanganese(I) is used in the formation of (eta6-arene)tricarbonylmanganese(I) by reacting with arene (arene= hexamethyl benzene, 1,2,4,5-tetramethyl benzene, mesitylene, p-xylene and toluene) in the presence silver salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14516-54-2
Molecular Formula C5BrMnO5-
Molecular Weight (g/mol) 274.892
MDL Number MFCD00049806
InChI Key LOCZKKJCQVRJPQ-UHFFFAOYSA-M
Synonym bromopentacarbonylmanganese i
PubChem CID 10978692
IUPAC Name carbon monoxide;manganese;bromide
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mn].[Br-]

Specifications

Color Orange
Linear Formula Mn(CO)5Br
Quantity 5 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Solubility Information Soluble in organic solvents.
Formula Weight 274.89
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Bromopentacarbonylmanganese(I)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.