missing translation for 'onlineSavingsMsg'
Learn More

Melford Bromophenol Blue

Product Code. 30018082
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018082 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018082 Supplier Melford Supplier No. B4016025.0
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Substance Name: 3',3',5',5'-Tetrabromo-phenolsulfonephthalein
  • CAS Number: 115-39-9
  • Molecular Formula: C19H10Br4O5S
  • Molecular Weight: 669.9

Specifications

Chemical Name or Material Bromophenol Blue
CAS 115-39-9
Molecular Formula C19H10Br4O5S
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.