missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue, for Electrophoresis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Specifications
Specifications
| Melting Point | 279°C |
| CAS | 115-39-9 |
| Molecular Formula | C19H10Br4O5S |
| MDL Number | 5875 |
| Formula Weight | 669.96g/mol |
| Packaging | Glass Jar |
| Color | Red to Violet |
| Quantity | 25 g |
| Physical Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction