missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, for Electrophoresis, Fisher Chemical™

Product Code. 10243420 Shop All Fisher Chemical Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10243420

Brand: Fisher Chemical B/P620/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 279°C
CAS 115-39-9
Molecular Formula C19H10Br4O5S
MDL Number 5875
Formula Weight 669.96g/mol
Packaging Glass Jar
Color Red to Violet
Quantity 5 g
Physical Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.