missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue (Free Acid), Fisher BioReagents

Product Code. 10679733
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10679733 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10679733 Supplier Fisher BioReagents Supplier No. BP11525
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

Glass Bottle; 25g

Bromophenol Blue is commonly used as a tracking dye in electrophoresis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromophenol Blue
Melting Point 279°C
Name Note Free Acid
CAS 115-39-9
Extinction Coefficient 1000g(-1) cm(-1) min.
Molecular Formula C19H10Br4O5S
Merck Index 15, 1454
Solubility Information Solubility in water: Soluble
Identification Pass Test
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
Molecular Weight (g/mol) 669.96
PubChem CID 8272
ChEBI CHEBI:59424
Lambda Max 589 to 593nm
Formula Weight 669.96
Loss on Drying 5% max.
Packaging Glass Bottle
Vapor Pressure Vapor Pressure: Negligible
Color Violet
Quantity 25 g
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.