missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, Indicator, Reag. Ph. Eur., Solstice

Product Code. 15613840 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15613840 5 g 5g
15603840 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15613840 Supplier Honeywell-Fluka Supplier No. 327125G
  • ONLINE EXCLUSIVE PRICE
    56.00 €
    47.45 € / 5g
    Save 8.55 €
    15% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 204°C
Packaging Glass Bottle
UN Number NONH for all modes of transport
Beilstein 61698
Color Red
pH 2.8 to 4.4
Quantity 5 g
Physical Form Powder
Formula Weight 669.96g/mol
Chemical Name or Material Bromophenol Blue, Reag. Ph. Eur.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.