missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, Pure, Indicator Grade, Solid, Fisher Chemical™

Product Code. 10233280 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10233280 25 g 25g
10223280 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10233280 Supplier Fisher Chemical Supplier No. B/4630/46
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Bromophenol Blue, Pure, Indicator Grade, Solid

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 279°C
CAS 115-39-9
Molecular Formula C19H10Br4O5S
MDL Number 5875
Formula Weight 669.96g/mol
Packaging Glass Jar
Color Red to Violet
Quantity 25 g
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.