missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Bromophenol Blue sodium salt, 0.04% w/v aq. soln.
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
6 x 100 mL
Förpackningsstorlek
100mL
500mL
600mL
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Concentration | 0.04% w/v aq. soln. |
| Merck Index | 141444 |
| Formula Weight | 691.96 |
| Color | Red |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Bromophenol Blue sodium salt |
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion