missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS reagent

Product Code. 10045593 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
Unit Size:
100g
10g
50g
5g
This item is not returnable. View return policy

Product Code. 10045593

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indicator | CAS: 34725-61-6 | C19H9Br4NaO5S | 691.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H9Br4NaO5S
Molecular Weight (g/mol) 691.94
MDL Number MFCD00013793
InChI Key KGANHHDZDYZJEM-UHFFFAOYNA-M
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein
PubChem CID 3791690
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate

Specifications

Melting Point >300.0°C
Beilstein 19, I, 649
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
Formula Weight 691.94
Grade ACS Reagent
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material Bromophenol Blue, sodium salt, Pure

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.