missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, Sodium Salt, MP Biomedicals™

Product Code. 11432802 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432802 5 g 5g
11481612 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432802 Supplier MP Biomedicals Supplier No. 0215250605
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Bromophenol Blue, Sodium Salt is a tracking dye.

  • Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
  • It is a tracking dye for alkaline and neutral buffer systems, as well as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym 3',3”,5',5”-tetrabromophenolsulfonphthalein sodium salt, 3', 3”, 5'

Specifications

CAS 62625-28-9
Molecular Formula C19H9Br4NaO5S
InChI Key TWKDJMRHSLWRFI-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol) 691.942
PubChem CID 3791690
Formula Weight 692
Color Green
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Bromophenol blue, sodium salt, For ACS analysis
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.