missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue Solution for PANPEHA indicator paper, Solstice™

Product Code. 17234546
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17234546 2.5 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17234546 Supplier Fluka Supplier No. 310682.5L
  • ONLINE EXCLUSIVE PRICE
    1675.00 €
    1512.15 € / 2.5 L
    Save 162.85 €
    9% Off
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Bromophenol blue sol
Melting Point -115°C (The physical data is that of the main component)
Boiling Point 78°C to 79°C (1013 hPa) (The physical data is that of the main component)
CAS 115-39-9, 64-17-5, 78-93-3
UN Number UN1170
Color Blue
Quantity 2.5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.