missing translation for 'onlineSavingsMsg'
Learn More

Bromosuccinic Acid, TRC

Product Code. 30331074
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30331074

Brand: Toronto Research Chemicals TRCB687800100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromosuccinic Acid
CAS 923-06-8
Molecular Formula C4H5BrO4
Quantity 100 g
Synonym 2-Bromobutanedioic Acid, Bromobutanedioic Acid, 2-Bromosuccinic Acid, DL-Bromosuccinic Acid, Monobromosuccinic Acid, NSC 227872, (±)-Bromosuccinic Acid
InChI Formula InChI=1S/C4H5BrO4/c5-2(4(8)9)1-3(6)7/h2H,1H2,(H,6,7)(H,8,9)
SMILES OC(=O)CC(Br)C(=O)O
IUPAC Name 2-bromobutanedioic acid
Molecular Weight (g/mol) 196.98
Formula Weight 195.9371
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.