missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromosuccinic Acid, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Bromosuccinic Acid |
| CAS | 923-06-8 |
| Molecular Formula | C4H5BrO4 |
| Quantity | 25 g |
| Synonym | 2-Bromobutanedioic Acid, Bromobutanedioic Acid, 2-Bromosuccinic Acid, DL-Bromosuccinic Acid, Monobromosuccinic Acid, NSC 227872, (±)-Bromosuccinic Acid |
| InChI Formula | InChI=1S/C4H5BrO4/c5-2(4(8)9)1-3(6)7/h2H,1H2,(H,6,7)(H,8,9) |
| SMILES | OC(=O)CC(Br)C(=O)O |
| IUPAC Name | 2-bromobutanedioic acid |
| Molecular Weight (g/mol) | 196.98 |
| Formula Weight | 195.9371 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction