missing translation for 'onlineSavingsMsg'
Learn More

Bromosulfalein, TRC

Product Code. 30347774
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347774 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347774 Supplier Toronto Research Chemicals Supplier No. TRCB6891001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromosulfalein
CAS 71-67-0
Molecular Formula C20 H8 Br4 O10 S2 . 2 Na
Quantity 1 g
Synonym Benzenesulfonic acid, 3,3'-(4,5,6,7-tetrabromo-3-oxo-1(3H)-isobenzofuranylidene)bis[6-hydroxy-, disodium salt, Phenolphthalein, 4,5,6,7-tetrabromo-3',3''-disulfo-, disodium salt (8CI), BSF, BSF Simes, BSP, BSP (pharmaceutical), BSP sodium, Brom-tetragnost, Bromosulfalein, Bromosulfophthalein, Bromosulphalein, Bromosulphthalein, Bromotaleina, Bromsulfalein, Bromsulfan, Bromsulfophthalein, Bromsulfthalein, Bromsulphalein, Bromsulphthalein, Bromthalein, CBSP, Disodium bromosulfophthalein, Hepartest, Hepatestabrome, Hepatosulfalein, Phenoltetrabromophthaleinsulfonate, Sodium bromosulfalein, Sodium bromosulfophthalein, Sodium bromsulphalein, Sodium bromsulphthalein, Sodium phenol tetrabromophthalein, Sodium sulfobromophthalein, Sodium sulphobromophthalein, Sulfobromophthalein, Sulfobromophthalein sodium, Sulfobromphthalein, Sulphobromophthalein, Sulphobromophthalein sodium, Tetrabromosulfophthalein, Tetrabromsulfthalein
InChI Formula InChI=1S/C20H10Br4O10S2.2Na/c21-15-13-14(16(22)18(24)17(15)23)20(34-19(13)27,7-1-3-9(25)11(5-7)35(28,29)30)8-2-4-10(26)12(6-8)36(31,32)33;;/h1-6,25-26H,(H,28,29,30)(H,31,32,33);;/q;2*+1/p-2
SMILES [Na+].[Na+].Oc1ccc(cc1S(=O)(=O)[O-])C2(OC(=O)c3c(Br)c(Br)c(Br)c(Br)c23)c4ccc(O)c(c4)S(=O)(=O)[O-]
IUPAC Name disodium;2-hydroxy-5-[4,5,6,7-tetrabromo-1-(4-hydroxy-3-sulfonatophenyl)-3-oxo-2-benzofuran-1-yl]benzenesulfonate
Molecular Weight (g/mol) 838
Formula Weight 833.6088
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.