missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Sultone Form, ACS Reagent, Reag. Ph. Eur., Solstice

Product Code. 15643840
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
Unit Size:
1kg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15643840 25 g Glass Bottle 25g
15653840 5 g Glass Bottle 5g
15633840 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15643840 Supplier Honeywell-Fluka Supplier No. 3271425G
  • ONLINE EXCLUSIVE PRICE
    144.00 €
    120.41 € / 25g
    Save 23.59 €
    16% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator, ACS Reagent, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym 3′,3″-Dibromothymolsulfonphthalein
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

Packaging Glass Bottle
Suitability Complies for clarity of solution
Visual Transition Interval 5.8 to 7.4
UN Number NONH for all modes of transport
Beilstein 373934
Color Violet
Quantity 25 g
Physical Form Powder
Formula Weight 624.38g/mol
Grade ACS Reagent
Chemical Name or Material Bromothymol Blue Sultone Form, Reag. Ph. Eur.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.