missing translation for 'onlineSavingsMsg'
Learn More

Bromotrimethylsilane, 97%, stab. with copper powder or silver wire

Product Code. 11408246
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408246 250 g 250g
11488236 10 g 10g
11498236 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11408246 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. A15334.30
 Restricted Item
Estimated Shipment: 30-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 2857-97-8 | C3H9BrSi | 153.094 g/mol

Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.

Solubility
Miscible with carbon tetrachloride, chloroform, dichloroethane, acetonitrile, toluene, dichloromethane and hexane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids, water and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 2857-97-8
Molecular Formula C3H9BrSi
Molecular Weight (g/mol) 153.094
MDL Number MFCD00000048
InChI Key IYYIVELXUANFED-UHFFFAOYSA-N
Synonym trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
PubChem CID 76113
IUPAC Name bromo(trimethyl)silane
SMILES C[Si](C)(C)Br

Spezifikation

Boiling Point 78°C to 80°C
Melting Point -44°C
Quantity 250 g
Flash Point −3°C (26°F)
Odor Acrid
Refractive Index 1.425
Sensitivity Moisture sensitive
Density 1.183
Linear Formula (CH3)3SiBr
UN Number UN2924
Beilstein 1731135
Solubility Information Miscible with carbon tetrachloride,chloroform,dichloroethane,acetonitrile,toluene,dichloromethane and hexane.
Formula Weight 153.09
Percent Purity 97%
Chemical Name or Material Bromotrimethylsilane, stabilized with copper powder or silver wire
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.